Learn More
Sartorius™ Cylindrical Calibration Weight
Product Code. 11363424 Shop All Sartorius Products
Click to view available options
Mass:
200 g
2 kg
Unit Size:
200g
Each
Product Code. 11363424
Brand: Sartorius™ YCW624AC00
Only null left
Add to basket
Add to basket
Confidently calibrate your scale with this finely crafted, special steel weight. Sartorius™ Cylindrical Calibration Weight is highly polished and non-magnetizable.
  • Cylindrical weight
  • Special steel
  • Highly polished
  • Non-magnetizable
  • Highly corrosion-resistant
  • Class F2 - suitable for use for traceability of OIML class M1 weights and lower
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Weight
Classification Class F2 - suitable for use for traceability of OIML class M1 weights and lower
Material Special Steel
Case Material Plastic
Density 7,950kg/m3
Mass 2 kg
For Use With (Equipment) Laboratory scale