Learn More
Sartorius™ Cylindrical Calibration Weight
Shop All Sartorius Products
Click to view available options
Mass:
200 g
2 kg
Unit Size:
200g
Each
Description
- Cylindrical weight
- Special steel
- Highly polished
- Non-magnetizable
- Highly corrosion-resistant
- Class F2 - suitable for use for traceability of OIML class M1 weights and lower
Specifications
Specifications
| Product Type | Calibration Weight |
| Classification | Class F2 - suitable for use for traceability of OIML class M1 weights and lower |
| Material | Special Steel |
| Case Material | Plastic |
| Density | 7,950kg/m3 |
| Mass | 200 g |
| For Use With (Equipment) | Laboratory scale |