Learn More
Kartell™ Polypropylene Centrifuge Tubes
Manufactured from highly translucent polypropylene for  enhanced visibility and good chemical resistance. Autoclavable, rimmed Kartell™ Centrifuge Tubes deliver stability in a properly balanced rotor and are suitable for use as a conventional test tube.
Brand:  Kartell™ 0030200
Product Code. 10386011
  • € 47.40 / Pack of 100
Only null left
Estimated Shipment: 10-02-2026
Description
Manufactured from highly translucent polypropylene for  enhanced visibility and good chemical resistance. Autoclavable, rimmed Kartell™ Centrifuge Tubes deliver stability in a properly balanced rotor and are suitable for use as a conventional test tube.
  • Solid polypropylene construction translates to safety and economy with reduced breakage in the lab
  • Withstands temperatures up to 120°C
  • Rimmed
  • Autoclavable
  • Doubles as a laboratory test tube*
*Cannot be used with naked flame