Learn More
Fisherbrand™ Cylindrical PTFE or Glass Stir Bars
Alnico V magnet core encapsulated by PTFE or glass. Fisherbrand™ Cylindrical PTFE or Glass Stir Bars are autoclavable.
Brand: Fisherbrand™
Additional Details : Weight : 0.04512kg
Description
- Stable, general-purpose magnetic stir bar
- Smooth, round profile for a wide variety of applications
- Autoclavable