Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessel, Round Bottom
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3 L
4 L
5 L
6 L
10 L
20 L
Includes:
Laboratory Flange 100
Laboratory Flange 150
Laboratory Flange 200
Laboratory Flange 60
Unit Size:
Each
Specifications
Specifications
| Description | DURAN™ Tubing |
| Capacity (Metric) | 4 L |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 335 mm |
| Includes | Laboratory Flange 150 |
| Product Type | Reaction vessel |
| Shape | Cylindrical |
| Bottom Shape | Round |
| Diameter (Metric) Outer | 160 mm |