missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CYM 5520
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CYM 5520 is a selective S1P2 allosteric agonist (EC50 = 0.48 μM). Selective for S1P2 receptors over a panel of 29 other receptors and transporters.
Specifications
Specifications
| Chemical Name or Material | 1-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethyl]-1,6-dihydro-6-oxo-3-pyridinecarbonitrile |
| CAS | 1449747-00-5 |
| Quantity | 50 mg |
| Target | Sphingosine-1-phosphate Receptor Agonists |
| Molecular Formula | C21H19N3O2 |
| Purity | 0.99 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction