missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 9484
GREENER_CHOICE

Product Code. 30001533
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001533

Brand: Tocris Bioscience™ 4606/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent NPY Y2 receptor antagonist

Potent neuropeptide Y (NPY) Y2 receptor antagonist (IC50 = 19 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1383478-94-1
Molecular Formula C27H31N3O3S2

Specifications

Quantity 50 mg
Formula Weight 509.63
Purity >98%
Molecular Weight (g/mol) 509.683
Chemical Name or Material CYM 9484
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.