missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP11B2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | CYP11B2 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CYP11B2 Polyclonal antibody specifically detects CYP11B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CYP11B2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 1585 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ALDOSCYP11B, Aldosterone synthase, Aldosterone-synthesizing enzyme, CPN2, CYP11BL, CYPXIB2, cytochrome P450 11B2, mitochondrial, cytochrome P450, family 11, subfamily B, polypeptide 2, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2, Cytochrome P-450Aldo, Cytochrome P-450C18, EC 1.14.15, EC 1.14.15.4, EC 1.14.15.5, mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2, P450aldo, P450C18, P-450C18, steroid 11-beta/18-hydroxylase, steroid 11-beta-monooxygenase, Steroid 18-hydroxylase, steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 234-503 of human CYP11B2 (NP_000489.3). TVQLMFMPRSLSRWISPKVWKEHFEAWDCIFQYGDNCIQKIYQELAFNRPQHYTGIVAELLLKAELSLEAIKANSMELTAGSVDTTAFPLLMTLFELARNPDVQQILRQESLAAAASISEHPQKATTELPLLRAALKETLRLYPVGLFLERVVSSDLVLQNYHIPAGTLVQVFLYSLGRNAALFPRPERYNPQRWLDIRGSGRNFHHVPFGFGMRQCLGRRLAEAEMLLLLHHVLKHFLVETLTQEDIKMVYSFILRPGTSPLLTFRAIN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title