missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1A2 Monoclonal Antibody (OTI2E9), TrueMAB™, OriGene
Description
CYP1A2 Monoclonal antibody specifically detects CYP1A2 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
| Antigen | CYP1A2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI2E9 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Cyp1a2 |
| Gene Accession No. | P05177 |
| Gene Alias | aromatic compound inducible; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP12; CYP; Cyp1a1; Cyp1a2; Cyp1a-2; CYPD45; CYPIA2; Cytochrome P(3)450; cytochrome P-448; cytochrome P450 1A2; Cytochrome P450 4; cytochrome P450 family 1 subfamily A member 2; cytochrome P450 family 1 subfamily a polypeptide 1; Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily A, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; Cytochrome P-450d; cytochrome P450-D; Cytochrome P450-P2; cytochrome P450-P3; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P(3)450; P3-450; P-448; P450; P450 4; P450 form 4; P450(PA); P450-3; P-450d; P450-D; P450-P3; RATCYPD45; xenobiotic monooxygenase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?