missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Antibody (5E2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50190AF750
This item is not returnable.
View return policy
Description
CYP1B1 Monoclonal specifically detects CYP1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CYP1B1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CYP1B1 | |
| Ovalbumin-conjugated synthetic peptide - PENFDPARFLDKDGL | |
| 0.1 mL | |
| Primary | |
| Does not react with CYP1A1. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry | |
| 500 | |
| Western Blot, ELISA, Immunohistochemistry | |
| aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase | |
| Mouse | |
| Protein A purified | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1545.0 | |
| Human, Mouse, Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction