missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP20A1 Antibody (Z27-P3H7*E3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50192G
This item is not returnable.
View return policy
Description
CYP20A1 Monoclonal specifically detects CYP20A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CYP20A1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP20A1 | |
| Ovalbumin-conjugated synthetic peptide AWITVSKRY | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 57404 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Z27-P3H7*E3 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CYP-M, cytochrome P450 20A1, cytochrome P450 monooxygenase, cytochrome P450, family 20, subfamily A, polypeptide 1, EC 1.14, MGC22229 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction