missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26A1 Antibody (F27P6A1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50193AF405
This item is not returnable.
View return policy
Description
CYP26A1 Monoclonal specifically detects CYP26A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CYP26A1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP26A1 | |
| Ovalbumin-conjugated synthetic peptide PARFTHFHGE (C-terminal sequence) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1592 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry | |
| F27P6A1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction