missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP26A1 Polyclonal Antibody

Product Code. 17793444
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17793444 20 μL 20µL
17790474 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17793444 Supplier Invitrogen™ Supplier No. PA5122051

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP26A1 Polyclonal Antibody for Western Blot, IHC (P)

CYP26A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein acts on retinoids, including all-trans-retinoic acid (RA), with both 4-hydroxylation and 18-hydroxylation activities. This enzyme regulates the cellular level of retinoic acid which is involved in regulation of gene expression in both embryonic and adult tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP26A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CYP26A1
Gene Accession No. O43174
Gene Alias CP26; Cyp26; CYP26A1; Cytochrome P450 26A1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 retinoic acid-inactivating 1; cytochrome P450, 26, retinoic acid; cytochrome P450, 26, retinoic acid A1; cytochrome P450, family 26, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIA, polypeptide 1; cytochrome P450RAI; hP450RAI; P450, retinoic acid-inactivating, 1; P450RA; P450RAI; P450RAI1; RAH; Retinoic acid 4-hydroxylase; retinoic acid hydrolase; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome
Gene Symbols CYP26A1
Host Species Rabbit
Immunogen Recombinant protein Cytochrome P450 26A1. The antigen corresponds to amino acid range 73-255 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100124374, 1592
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.