missing translation for 'onlineSavingsMsg'
Learn More

CYP2A13 Antibody, Novus Biologicals™

Product Code. 18622798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18622798 100 μg 100µL
18661448 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18622798 Supplier Novus Biologicals Supplier No. NBP306099100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP2A13 Polyclonal antibody specifically detects CYP2A13 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2A13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, pH7.0
Gene Alias CPAD, CYP2A, CYPIIA13, cytochrome P450 2A13, cytochrome P450, family 2, subfamily A, polypeptide 13, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 13, EC 1.14.14.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYP2A13 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1553
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.