missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2A6 Monoclonal Antibody (OTI5F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503805
This item is not returnable.
View return policy
Description
This gene, CYP2A6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to hydroxylate coumarin, and also metabolizes nicotine, aflatoxin B1, nitrosamines, and some pharmaceuticals. Individuals with certain allelic variants are said to have a poor metabolizer phenotype, meaning they do not efficiently metabolize coumarin or nicotine. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. The gene was formerly referred to as CYP2A3; however, it has been renamed CYP2A6.Specifications
| CYP2A6 | |
| Monoclonal | |
| 0.83 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11509 | |
| CYP2A6 | |
| Full length human recombit protein of human CYP2A6 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F8 | |
| Unconjugated | |
| CYP2A6 | |
| CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1548 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction