missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2A6 Monoclonal Antibody (OTI9G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504099S
This item is not returnable.
View return policy
Description
CYP2A6 Monoclonal antibody specifically detects CYP2A6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CYP2A6 | |
| Monoclonal | |
| 1.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01036 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1472 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9G3 | |
| Unconjugated | |
| CST4 | |
| 1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CYP; CYP2A; CYP2A3; CYP2A6; CYPIIA6; cytochrome P450 2A6; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 IIA3; Cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; flavoprotein-linked monooxygenase; P450; P450 IIA3; P450(I); P450C2A; P450PB; xenobiotic monooxygenase | |
| Full length human recombinant protein of human CST4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction