missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyp2e1, Mouse anti-Rat, Clone: 1-98-1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB3817.100uL
This item is not returnable.
View return policy
Specifications
| Cyp2e1 | |
| Monoclonal | |
| Unconjugated | |
| In purified ascites | |
| Cyp2e | |
| Mouse | |
| 100 μL | |
| Primary | |
| This mouse monoclonal antibody is specific to rat cytochrome P450 2E1. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot | |
| 1-98-1 | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| cytochrome P450, family 2, subfamily e, polypeptide 1 | |
| Cyp2e1 | |
| Recombinant protein corresponding to rat Cyp2e1. | |
| RUO | |
| 25086 | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction