Learn More
Invitrogen™ CYP2E1 Recombinant Rabbit Monoclonal Antibody (5G8P6)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557857
 View more versions of this product
Product Code. 30283565
  • € 176.00 / 20µL
Description
CYP2E1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CYP2E1
Recombinant Monoclonal
0.13 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P05181, P05182, Q05421
Cyp2e1
Recombinant protein of human Cytochrome P450 2E1 (CYP2E1).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot
5G8P6
Unconjugated
Cyp2e1
4-nitrophenol 2-hydroxylase; CP2E1; CPE1; Cyp2e; Cyp2e1; Cyp2e-1; CYPIIE1; cytochrome P450 2E1; cytochrome P450 family 2 subfamily E member 1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily e, polypeptide 1; cytochrome P450, subfamily 2E, polypeptide 1; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; cytochrome P450-ALC; Cytochrome P450-J; cytochrome P450RLM6; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450C2E; P450-J; P450RLM6; xenobiotic monooxygenase
Rabbit
Protein A
RUO
13106, 1571, 25086
-20°C, Avoid Freeze/Thaw Cycles