missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2J2 Monoclonal Antibody (OTI3B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503635S
This item is not returnable.
View return policy
Description
CYP2J2 Monoclonal antibody specifically detects CYP2J2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CYP2J2 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Albendazole monooxygenase (hydroxylating); Albendazole monooxygenase (sulfoxide-forming); Arachidonic acid epoxygenase; CPJ2; CYP2J2; Cyp2j4; CYPIIJ2; cytochrome P450 2J2; cytochrome P450 family 2 subfamily J member 2; cytochrome P450, family 2, subfamily J, polypeptide 2; cytochrome P450, family 2, subfamily j, polypeptide 4; cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate isomerase; microsomal monooxygenase | |
| Full length human recombinant protein of human KIAA0495 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B4 | |
| Unconjugated | |
| TP73-AS1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57212 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction