missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2J2 Monoclonal Antibody (OTI3B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503635
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is thought to be the predomit enzyme responsible for epoxidation of endogenous arachidonic acid in cardiac tissue.Specifications
| CYP2J2 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P51589 | |
| CYP2J2 | |
| Full length human recombit protein of human CYP2J2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B4 | |
| Unconjugated | |
| CYP2J2 | |
| CPJ2, CYPIIJ2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1573 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction