missing translation for 'onlineSavingsMsg'
Learn More

CYP2J2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16821844 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821844 20 μL 20µL
16811844 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16821844 Supplier Proteintech Supplier No. 135621AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is thought to be the predomit enzyme responsible for epoxidation of endogenous arachidonic acid in cardiac tissue.

Specifications

Antigen CYP2J2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP2J2
Gene Accession No. P51589
Gene Alias Arachidonic acid epoxygenase, CPJ2, CYP2J2, CYPIIJ2, Cytochrome P450 2J2
Gene Symbols CYP2J2, Cyp2j4
Host Species Rabbit
Immunogen CYP2J2 Fusion Protein Ag4435
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1573, 65210
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.