missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2R1 Antibody (M26-P6H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50204CL1
This item is not returnable.
View return policy
Description
CYP2R1 Monoclonal antibody specifically detects CYP2R1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| CYP2R1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cytochrome P450 2R1, cytochrome P450, family 2, R1, cytochrome P450, family 2, subfamily R, polypeptide 1, EC 1.14.13.15, MGC4663, vitamin D 25-hydroxylase | |
| Ovalbumin-conjugated synthetic peptide QPYLICAERR (C-terminal sequence). | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 120227 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 λ |
| ELISA, Immunohistochemistry, Western Blot | |
| M26-P6H1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction