missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP39A1 Antibody (M30P6D6), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50206MFV610
This item is not returnable.
View return policy
Description
CYP39A1 Monoclonal antibody specifically detects CYP39A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| CYP39A1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| M30P6D6 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Cytochrome P450 39A1, cytochrome P450, family 39, subfamily A, polypeptide 1, cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1,24-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.99, hCYP39A1, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase | |
| Ovalbumin-conjugated synthetic peptide CRIEYKQRI (C-terminal sequence). | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 51302 | |
| Store at 4C in the dark. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction