missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP39A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538909
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].
Specifications
| CYP39A1 | |
| Polyclonal | |
| Unconjugated | |
| CYP39A1 | |
| CYP39A1 | |
| A synthetic peptide derived from the internal region of human Cytochrome P450 39A1 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NYL5, Q9JKJ9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51302, 56050 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction