Learn More
Invitrogen™ CYP3A5 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA514903
Product Code. 10324250
  • € 518.00 / 400µL
Description
CYP3A5 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
CYP3A5 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. The enzyme metabolizes drugs such as nifedipine and cyclosporine as well as the steroid hormones testosterone, progesterone and androstenedione.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CYP3A5
Polyclonal
Unconjugated
CYP3A5
aryl hydrocarbon hydroxylase; CP35; CYP3A28; CYP3A4; CYP3A5; CYPIIIA28; CYPIIIA5; cytochrome P450; cytochrome P450 3A28; cytochrome P450 3A5; cytochrome P450 family 3 subfamily A member 5; cytochrome P450 HLp2; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; cytochrome P450-PCN3; flavoprotein-linked monooxygenase; HLp2; microsomal monooxygenase; P450PCN3; P450-PCN3; PCN3; Unknown (protein for MGC:127840); xenobiotic monooxygenase
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
1577
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P20815
CYP3A5
KLH conjugated synthetic peptide between 476-502 amino acids from the C-terminal region of human CYP3A5.
400 μL