missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP3C1 Antibody (V12P6F5*E3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50210F
This item is not returnable.
View return policy
Description
CYP3C1 Monoclonal specifically detects CYP3C1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| CYP3C1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| V12P6F5*E3 | |
| ELISA, Immunohistochemistry | |
| CYP3A, cyp3c1, Cytochrome P450, family 3, subfamily c, polypeptide 1, wu:fa96d12, wu:fc26b04, zgc:56626 | |
| Ovalbumin-conjugated synthetic peptide KLIPRSHKEK (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction