Learn More
CYP46A1 Mouse, anti-Human, Clone: 1E10D3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68576-1-IG-150UL
 View more versions of this product
Product Code. 18066205
  • € 410.00 / 150µL
Description
The CYP46A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP46A1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The CYP46A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CYP46A1
1E10D3
CH24H, Cholesterol 24 hydroxylase, CP46, CYP46, CYP46A1, Cytochrome P450 46A1
150 μL
10858
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunohistochemistry, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Rabbit, Pig
150 kDa
IgG2b