missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP4F12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113342
This item is not returnable.
View return policy
Description
CYP4F12 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CYP4F12 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein likely localizes to the endoplasmic reticulum.
Specifications
| CYP4F12 | |
| Polyclonal | |
| Unconjugated | |
| CYP4F12 | |
| CYP4F12; CYPIVF12; Cytochrome P450 4F12; cytochrome P450 family 4 subfamily F member 12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12; F22329_1; UNQ568/PRO1129 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66002 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HCS2 | |
| CYP4F12 | |
| Recombinant Human Cytochrome P450 4F12 protein (265-524AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction