Learn More
Invitrogen™ CYP4F12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596890
Description
Immunogen sequence: DFTDAVIRER RRTLPTQGID DFFKDKAKSK TLDFIDVLLL SKDEDGKALS DEDIRAEADT FMFGGHDTTA SGLSWVLYNL ARHPEYQERC RQEVQELLKD RDPKEIEWDD LAQLPFLTMC VKESLRLHPP APFISRCCTQ DIVLPDGRVI PKGITCLIDI IGVHHNPTVW PDPEVYDPFR FDPENSKGRS PLAFIPFSAG PRNCIGQAFA MAEMKVVLAL MLLHFRFLPD HTEPRRKLEL IMRAEGGLWL RVEPLNVSLQ; Positive Samples: Mouse placenta, Rat brain; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Multi-pass membrane protein.
CYP4F12 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein likely localizes to the endoplasmic reticulum.
Specifications
| CYP4F12 | |
| Polyclonal | |
| Unconjugated | |
| CYP4F12 | |
| CYP4F12; CYPIVF12; Cytochrome P450 4F12; cytochrome P450 family 4 subfamily F member 12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12; F22329_1; UNQ568/PRO1129 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66002 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HCS2 | |
| CYP4F12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-524 of human CYP4F12 (NP_0764333). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.