missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4F2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46854-25ul
This item is not returnable.
View return policy
Description
CYP4F2 Polyclonal antibody specifically detects CYP4F2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CYP4F2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q9HCS2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| CPF2, CYPIVF2, Cytochrome P450 4F2, cytochrome P450, family 4, subfamily F, polypeptide 2, cytochrome P450, subfamily IVF, polypeptide 2, Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega-hydroxylase, Leukotriene-B(4) 20-monooxygenase 1, leukotriene-B(4) omega-hydroxylase 1, leukotriene-B4 20-monooxygenase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KSKTLDFIDVLLLSKDEDGKALSDEDI | |
| 25 μL | |
| Lipid and Metabolism | |
| 8529 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction