Learn More
Invitrogen™ CYP4Z1/CYP4Z2P Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599832
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytochrome P450 4Z1/2.
The cytochrome P450 (P450) superfamily of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as nicotinamide adenine dinucleotide phosphate (NADPH) and P450 reductase. There are over 200 known genes which encode P450s. Epoxygenases are those P450s which metabolize AA to epoxyeicosatrienoic acid (EETs) and omega-hydroxylases are those P450s which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). As well as fatty acid metabolism, P450s also metabolize many drugs and toxins. Cytochrome P450 3A4 is abundantly expressed in liver and small intestine and is inducible by barbiturates, glucocorticoids and rifampicin.
Specifications
| CYP4Z1/CYP4Z2P | |
| Polyclonal | |
| Unconjugated | |
| CYP4Z1 | |
| CYP4A20; CYP4Z1; CYP4Z1/2; CYP4Z2P; CYPIVZ1; cytochrome P450 4Z1; cytochrome P450 family 4 subfamily Z member 1; cytochrome P450 family 4 subfamily Z member 2, pseudogene; cytochrome P450, family 4, subfamily Z, polypeptide 1; P450 4Z1/2; UNQ3060/PRO9882 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 163720, 199974 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86W10, Q8N1L4 | |
| CYP4Z1, CYP4Z2P | |
| A synthesized peptide derived from human CYP4Z1(Accession Q86W10), corresponding to amino acid residues S240-S290. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |