Learn More
CYP51A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13431-1-AP-150UL
 View more versions of this product
Product Code. 16851644
  • € 410.00 / 150µL
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein participates in the synthesis of cholesterol by catalyzing the removal of the 14alpha-methyl group from lanosterol. Homologous genes are found in all three eukaryotic phyla, fungi, plants, and animals, suggesting that this is one of the oldest cytochrome P450 genes. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CYP51A1
Polyclonal
Unconjugated
CYP51A1
CP51, CYP51, CYP51A1, CYPL1, CYPLI, Cytochrome P450 14DM, Cytochrome P450 51A1, Cytochrome P45014DM, Cytochrome P450LI, LDM, P450 14DM, P450L1, Sterol 14 alpha demethylase
Rabbit
Antigen Affinity Chromatography
RUO
13121, 1595, 25427
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16850, Q64654, Q8K0C4
Cyp51a1
CYP51A1 Fusion Protein Ag4235
150 μL
Primary
Human, Mouse, Rat