Learn More
CYP7B1 Antibody (M17-P3F2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50217-0.025ml
 View more versions of this product
Product Code. 18652488
  • € 201.00 / 0.03mL
Description
CYP7B1 Monoclonal antibody specifically detects CYP7B1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CYP7B1
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A
Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence)
0.025 mL
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience
9420
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 λ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
M17-P3F2
Unconjugated
PBS
Mouse
Protein A purified