missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP7B1 Antibody (M17-P3F2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50217UV
This item is not returnable.
View return policy
Description
CYP7B1 Monoclonal specifically detects CYP7B1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CYP7B1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP7B1 | |
| Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
| 9420 | |
| Store at 4C in the dark. | |
| IgG1 λ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| M17-P3F2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction