missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP8B1 Antibody (M15-P3B7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50218MFV500
This item is not returnable.
View return policy
Description
CYP8B1 Monoclonal antibody specifically detects CYP8B1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| CYP8B1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| M15-P3B7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CP8B, CYP127 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, CYPVIIIB1, Cytochrome P450 8B1, cytochrome P450, family 8, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1,7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase, EC 1.14.13.95, FLJ17826,7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, Sterol 12-alpha-hydroxylase | |
| Peptide sequence- QPSHDVRFR (amino acids 486 - 494) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1582 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction