missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystathionase Antibody (OTI1E12), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70488AF488
This item is not returnable.
View return policy
Description
Cystathionase Monoclonal specifically detects Cystathionase in Human, Mouse, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cystathionase | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CTH | |
| Full length recombinant protein of human Cystathionase (NP_001893) produced in HEK293T cells. | |
| 0.1 mL | |
| 1491.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1E12 | |
| Western Blot, Immunohistochemistry | |
| cystathionase (cystathionine gamma-lyase), cystathionine gamma-lyase, cysteine desulfhydrase, EC 4.4.1, EC 4.4.1.1, gamma-cystathionase, homoserine deaminase, homoserine dehydratase, MGC9471 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Canine, Monkey, Primate | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction