missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Antibody (236), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89524MFV610
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Western Blot
Specifications
| Cystatin A | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 236 | |
| Western Blot | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin A (Uniprot#: P01040; Ile2-Phe98). | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction