missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Antibody (CSTA/2882), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79882PCP
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Cystatin A | |
| Monoclonal | |
| PerCP | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| Recombinant fragment of human Cystatin A protein (around aa 1-98) (exact sequence is proprietary) (Uniprot: P01040) | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| CSTA/2882 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction