missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin C Antibody (033), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89501MFV500
This item is not returnable.
View return policy
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cystatin C | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 033 | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin C (Accession#: NP_000090.1; Ser27-Ala146). | |
| 0.1 mL | |
| Cancer, Epitope Tags, Signal Transduction | |
| 1471 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction