Learn More
Cystatin C Antibody (Cyst24cc) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NB100-62127JF525
Product Code. 30161580
  • € 507.00 / 0.20mL
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for ELISA,Immunoassay,Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin C
Monoclonal
Janelia Fluor 525
ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cystatin C purified from human urine.
0.2 mL
Cancer, Epitope Tags, Signal Transduction
1471
Store at 4°C in the dark.
IgG1
ELISA, Immunoassay, Sandwich ELISA
Cyst24cc
50mM Sodium Borate
Mouse
Protein A purified