missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin C Monoclonal Antibody (C9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541579
This item is not returnable.
View return policy
Description
Cystatin C Monoclonal Antibody for Western Blot
Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
Specifications
| Cystatin C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P01034 | |
| Cst3 | |
| Recombinant protein Cystatin C. The antigen corresponds to amino acid range 27-146 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| C9 | |
| Unconjugated | |
| Cst3 | |
| AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction