missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin D/Cst5 Antibody (011), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89539AF750
This item is not returnable.
View return policy
Description
Cystatin D/Cst5 Monoclonal antibody specifically detects Cystatin D/Cst5 in Human samples. It is validated for ELISA
Specifications
| Cystatin D/Cst5 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 011 | |
| ELISA | |
| cystatin 5, cystatin D, Cystatin-5, cystatin-D, cysteine-proteinase inhibitor, MGC71922 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin D/Cst5 (NP_001891.2; Met1-Val142). | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 1473 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction