missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Antibody (034), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89500FR
This item is not returnable.
View return policy
Description
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Human samples. It is validated for ELISA
Specifications
| Cystatin E/M/CST6 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 034 | |
| ELISA | |
| cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin E/M/CST6 (NP_001314.1; Met 1-Met 149). | |
| 0.1 mL | |
| Cancer | |
| 1474 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction