missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Antibody (MM0231-4H54), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12251AF647
This item is not returnable.
View return policy
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cystatin E/M/CST6 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q15828 | |
| CST6 | |
| Human recombinant Cystatin E/M/CST6 | |
| 0.1 mL | |
| Cancer | |
| 1474 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0231-4H54 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction