missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Antibody (RM0217-6F49), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12252F
This item is not returnable.
View return policy
Description
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot, Neutralization
Specifications
| Cystatin E/M/CST6 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Neutralization | |
| RM0217-6F49 | |
| Western Blot, Block/Neutralize | |
| cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
| Mouse recombinant Cystatin E/M/CST6 | |
| 0.1 mL | |
| Cancer | |
| 1474 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction