missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16074
This item is not returnable.
View return policy
Description
Cystatin F Polyclonal specifically detects Cystatin F in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cystatin F | |
| Polyclonal | |
| Unconjugated | |
| 1XPBS, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
| CST7 | |
| Recombinant protein encompassing a sequence within the center region of human Cystatin F. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 8530 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
| Rabbit | |
| 19 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction