missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin S Antibody (OTI2H10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70481H
This item is not returnable.
View return policy
Description
Cystatin S Monoclonal specifically detects Cystatin S in Human, Monkey samples. It is validated for Western Blot.
Specifications
| Cystatin S | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CST4 | |
| Full length recombinant protein of human Cystatin S (NP_001890) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| OTI2H10 | |
| Western Blot | |
| cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1 | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 1472.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction