Learn More
Invitrogen™ Cystatin S Monoclonal Antibody (OTI2H10)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525828
Product Code. 15985424
  • € 586.00 / 100µL
Description
Cystatin S Monoclonal Antibody for Western Blot, ICC/IF, Flow
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin S
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P01036
CST4
Full length human recombinant protein of CST4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot, Immunocytochemistry
OTI2H10
Unconjugated
CST4
CST4; cystatin 4; cystatin S; cystatin-4; Cystatin-S; cystatin-SA-III; Salivary acidic protein 1
Mouse
Affinity Chromatography
RUO
1472
-20°C, Avoid Freeze/Thaw Cycles