Learn More
Cystatin SA Antibody (005), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89916PE
Product Code. 18613101
  • € 607.00 / 0.10mL
Description
Cystatin SA Monoclonal antibody specifically detects Cystatin SA in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PE
ELISA, Sandwich ELISA Capture
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor
Store at 4C in the dark.