Learn More
Cystatin SA Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99787JF646
Product Code. 18458569
  • € 407.00 / 0.10mL
Description
Cystatin SA Polyclonal antibody specifically detects Cystatin SA in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin SA
50mM Sodium Borate
Purified
Polyclonal
Store at 4C in the dark.